Assay Details
| Gene Symbol: LOC4325640 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015793113.2 | XP_015648599.1 | 16 - 17 | 2124 | 110 | |
| GenBank mRNA | AK060447.1 | - | 10 - 11 | 1250 | 110 | |
| AK103297.1 | - | 15 - 16 | 2175 | 110 | ||
More Information
Set Membership: |
|

LOC4325640
Genomic Map