Assay Details
| Gene Symbol: LOC4325640 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015793113.2 | XP_015648599.1 | 3 - 4 | 440 | 88 | |
| GenBank mRNA | AK103297.1 | - | 3 - 4 | 408 | 88 | |
More Information
Set Membership: |
|

LOC4325640
Genomic Map