Assay Details
| Gene Symbol: LOC4325640 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015793113.2 | XP_015648599.1 | 9 - 10 | 1370 | 137 | |
| GenBank mRNA | AK103297.1 | - | 9 - 10 | 1337 | 137 | |
More Information
Set Membership: |
|

LOC4325640
Genomic Map