Assay Details
| Gene Symbol: LOC4324731 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015787164.2 | XP_015642650.1 | 1 - 2 | 421 | 67 | |
| GenBank mRNA | AY177699.1 | - | 1 - 2 | 186 | 67 | |
More Information
Set Membership: |
|

LOC4324731
Genomic Map