Assay Details
| Gene Symbol: LOC4324731 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015787164.2 | XP_015642650.1 | 5 - 6 | 738 | 107 | |
| GenBank mRNA | AY177699.1 | - | 5 - 6 | 503 | 107 | |
More Information
Set Membership: |
|

LOC4324731
Genomic Map