Assay Details
| Gene Symbol: LOC4327019 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015766213.2 | XP_015621699.1 | 2 - 3 | 864 | 67 | |
| XM_015766215.2 | XP_015621701.1 | 2 - 3 | 561 | 67 | ||
| GenBank mRNA | AK122112.1 | - | 2 - 3 | 854 | 67 | |
More Information
Set Membership: |
|

LOC4327019
Genomic Map