Assay Details
| Gene Symbol: LOC4327059 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015770217.2 | XP_015625703.1 | 1 | 876 | 86 | |
| GenBank mRNA | AK109278.1 | - | 1 | 450 | 86 | |
More Information
Set Membership: |
|

LOC4327059
Genomic Map