Assay Details
| Gene Symbol: LOC4325624 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015766515.2 | XP_015622001.1 | 6 - 7 | 1181 | 79 | |
| GenBank mRNA | AF200467.1 | - | 6 - 7 | 1133 | 79 | |
| AK243517.1 | - | 6 - 7 | 1147 | 79 | ||
More Information
Set Membership: |
|

LOC4325624
Genomic Map