Assay Details
| Gene Symbol: LOC4325091 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015761376.2 | XP_015616862.1 | 1 - 2 | 168 | 111 | |
| GenBank mRNA | AK069324.1 | - | 1 - 2 | 75 | 111 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015761376.2 transcripts.
Set Membership: |
|

LOC4325091
Genomic Map