Assay Details
| Gene Symbol: LOC4325091 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015761376.2 | XP_015616862.1 | 2 - 3 | 2737 | 88 | |
| GenBank mRNA | AK069324.1 | - | 2 - 3 | 2644 | 88 | |
More Information
Set Membership: |
|

LOC4325091
Genomic Map