Assay Details
| Gene Symbol: LOC4324867 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015760882.2 | XP_015616368.1 | 1 - 2 | 158 | 90 | |
| XM_015760892.2 | XP_015616378.1 | 1 - 2 | 157 | 90 | ||
| GenBank mRNA | AK099909.1 | - | 1 - 2 | 125 | 90 | |
| AK119276.1 | - | 1 - 2 | 124 | 90 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015760882.2, XM_015760892.2 transcripts.
Set Membership: |
|

LOC4324867
Genomic Map