Assay Details
| Gene Symbol: LOC4324867 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015760882.2 | XP_015616368.1 | 3 - 4 | 289 | 128 | |
| XM_015760892.2 | XP_015616378.1 | 3 - 4 | 288 | 128 | ||
| GenBank mRNA | AK069641.1 | - | 3 - 4 | 284 | 128 | |
| AK099909.1 | - | 3 - 4 | 256 | 128 | ||
| AK119276.1 | - | 3 - 4 | 255 | 128 | ||
More Information
Set Membership: |
|

LOC4324867
Genomic Map