Assay Details
| Gene Symbol: LOC4324867 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015760892.2 | XP_015616378.1 | 5 - 6 | 546 | 126 | |
| GenBank mRNA | AK069641.1 | - | 5 - 6 | 542 | 126 | |
| AK119276.1 | - | 5 - 6 | 513 | 126 | ||
More Information
Set Membership: |
|

LOC4324867
Genomic Map