Assay Details
| Gene Symbol: LOC4324867 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015760882.2 | XP_015616368.1 | 9 - 10 | 1123 | 66 | |
| XM_015760892.2 | XP_015616378.1 | 9 - 10 | 1095 | 66 | ||
| GenBank mRNA | AK069641.1 | - | 9 - 10 | 1091 | 66 | |
| AK099909.1 | - | 8 - 9 | 4722 | 66 | ||
More Information
Set Membership: |
|

LOC4324867
Genomic Map