Assay Details
| Gene Symbol: LOC4327748 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015765388.1 | XP_015620874.1 | 16 - 17 | 2052 | 109 | |
| XM_015765389.2 | XP_015620875.1 | 16 - 17 | 1674 | 109 | ||
| GenBank mRNA | AK289185.1 | - | 16 - 17 | 2064 | 109 | |
More Information
Set Membership: |
|

LOC4327748
Genomic Map