Assay Details
| Gene Symbol: LOC4323839 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015785189.1 | XP_015640675.1 | 4 - 5 | 751 | 82 | |
| GenBank mRNA | AK288993.1 | - | 4 - 5 | 675 | 82 | |
More Information
Set Membership: |
|

LOC4323839
Genomic Map