Assay Details
| Gene Symbol: LOC4328079 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015770879.2 | XP_015626365.1 | 2 - 3 | 718 | 120 | |
| GenBank mRNA | AF093628.1 | - | 2 - 3 | 224 | 120 | |
| AK242790.1 | - | 2 - 3 | 481 | 120 | ||
More Information
Set Membership: |
|

LOC4328079
Genomic Map