Assay Details
| Gene Symbol: LOC4328079 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015770879.2 | XP_015626365.1 | 4 - 5 | 974 | 103 | |
| GenBank mRNA | AF093628.1 | - | 4 - 5 | 480 | 103 | |
| AK242790.1 | - | 4 - 5 | 737 | 103 | ||
More Information
Set Membership: |
|

LOC4328079
Genomic Map