Assay Details
| Gene Symbol: LOC4328079 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015770879.2 | XP_015626365.1 | 5 - 6 | 1069 | 138 | |
| GenBank mRNA | AF093628.1 | - | 5 - 6 | 575 | 138 | |
| AK242790.1 | - | 5 - 6 | 832 | 138 | ||
More Information
Set Membership: |
|

LOC4328079
Genomic Map