Assay Details
| Gene Symbol: LOC4328149 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015770214.2 | XP_015625700.1 | 2 - 3 | 410 | 66 | |
| GenBank mRNA | AK061564.1 | - | 2 - 3 | 287 | 66 | |
| AK061953.1 | - | 2 - 3 | 398 | 66 | ||
| AK101060.1 | - | 2 - 3 | 409 | 66 | ||
More Information
Set Membership: |
|

LOC4328149
Genomic Map