Assay Details
| Gene Symbol: LOC4328208 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015771225.2 | XP_015626711.1 | 11 - 12 | 1630 | 131 | |
| XM_015771226.2 | XP_015626712.1 | 11 - 12 | 1599 | 131 | ||
| GenBank mRNA | AK060180.1 | - | 7 - 8 | 639 | 131 | |
More Information
Set Membership: |
|

LOC4328208
Genomic Map