Assay Details
| Gene Symbol: LOC4328508 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015768148.2 | XP_015623634.1 | 2 - 3 | 380 | 71 | |
| GenBank mRNA | AK101220.1 | - | 2 - 3 | 351 | 71 | |
More Information
Set Membership: |
|

LOC4328508
Genomic Map