Assay Details
| Gene Symbol: LOC4328597 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015769831.2 | XP_015625317.1 | 5 - 6 | 1414 | 83 | |
| GenBank mRNA | AK064096.1 | - | 5 - 6 | 1011 | 83 | |
More Information
Set Membership: |
|

LOC4328597
Genomic Map