Assay Details
| Gene Symbol: LOC4328779 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015768146.2 | XP_015623632.1 | 2 - 3 | 482 | 68 | |
| GenBank mRNA | AK121976.1 | - | 2 - 3 | 484 | 68 | |
More Information
Set Membership: |
|

LOC4328779
Genomic Map