Assay Details
| Gene Symbol: LOC4328814 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015767665.1 | XP_015623151.1 | 2 - 3 | 766 | 100 | |
| GenBank mRNA | AB856419.1 | - | 2 - 3 | 571 | 100 | |
| AK068806.1 | - | 2 - 3 | 667 | 100 | ||
More Information
Set Membership: |
|

LOC4328814
Genomic Map