Assay Details
| Gene Symbol: LOC4329094 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015768438.2 | XP_015623924.1 | 2 - 3 | 854 | 123 | |
| XM_015768439.2 | XP_015623925.1 | 2 - 3 | 854 | 123 | ||
| GenBank mRNA | AK121646.1 | - | 2 - 3 | 853 | 123 | |
More Information
Set Membership: |
|

LOC4329094
Genomic Map