Assay Details
| Gene Symbol: LOC4329137 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015771118.2 | XP_015626604.1 | 3 - 4 | 890 | 75 | |
| XM_015771119.2 | XP_015626605.1 | 3 - 4 | 241 | 75 | ||
| GenBank mRNA | AK072366.1 | - | 3 - 4 | 2033 | 75 | |
| AK101258.1 | - | 3 - 4 | 474 | 75 | ||
More Information
Set Membership: |
|

LOC4329137
Genomic Map