Assay Details
| Gene Symbol: LOC4329137 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015771118.2 | XP_015626604.1 | 9 - 10 | 1445 | 78 | |
| XM_015771119.2 | XP_015626605.1 | 9 - 10 | 796 | 78 | ||
| GenBank mRNA | AK072366.1 | - | 9 - 10 | 2605 | 78 | |
| AK101258.1 | - | 9 - 10 | 1029 | 78 | ||
More Information
Set Membership: |
|

LOC4329137
Genomic Map