Assay Details
| Gene Symbol: LOC4329192 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015772115.2 | XP_015627601.1 | 2 - 3 | 550 | 84 | |
| GenBank mRNA | AK073631.1 | - | 2 - 3 | 399 | 84 | |
| AK104165.1 | - | 2 - 3 | 313 | 84 | ||
More Information
Set Membership: |
|

LOC4329192
Genomic Map