Assay Details
| Gene Symbol: LOC4329234 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015771614.2 | XP_015627100.1 | 4 - 5 | 1049 | 87 | |
| GenBank mRNA | AK062153.1 | - | 3 - 4 | 396 | 87 | |
| AK067100.1 | - | 4 - 5 | 1040 | 87 | ||
More Information
Set Membership: |
|

LOC4329234
Genomic Map