Assay Details
| Gene Symbol: LOC4329560 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015769780.2 | XP_015625266.1 | 3 - 4 | 352 | 78 | |
| GenBank mRNA | AB062681.1 | - | 3 - 4 | 289 | 78 | |
| AK061722.1 | - | 3 - 4 | 309 | 78 | ||
| AK106147.1 | - | 3 - 4 | 309 | 78 | ||
| AK121892.1 | - | 3 - 4 | 358 | 78 | ||
More Information
Set Membership: |
|

LOC4329560
Genomic Map