Assay Details
| Gene Symbol: LOC4329631 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015771918.2 | XP_015627404.1 | 5 - 6 | 1100 | 147 | |
| GenBank mRNA | AJ428900.1 | - | 4 - 5 | 771 | 147 | |
| AK068448.1 | - | 5 - 6 | 1042 | 147 | ||
More Information
Set Membership: |
|

LOC4329631
Genomic Map