Assay Details
| Gene Symbol: LOC4329813 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015768325.2 | XP_015623811.1 | 1 - 2 | 375 | 121 | |
| GenBank mRNA | AK062460.1 | - | 1 - 2 | 205 | 121 | |
| GQ848055.1 | - | 1 - 2 | 185 | 121 | ||
More Information
Set Membership: |
|

LOC4329813
Genomic Map