Assay Details
| Gene Symbol: LOC4330553 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015767280.2 | XP_015622766.1 | 3 - 4 | 1393 | 72 | |
| XM_015767281.2 | XP_015622767.1 | 3 - 4 | 701 | 72 | ||
| GenBank mRNA | AK101764.1 | - | 3 - 4 | 1095 | 72 | |
More Information
Set Membership: |
|

LOC4330553
Genomic Map