Assay Details
| Gene Symbol: LOC4330553 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015767280.2 | XP_015622766.1 | 5 - 6 | 1958 | 90 | |
| XM_015767281.2 | XP_015622767.1 | 5 - 6 | 1266 | 90 | ||
| GenBank mRNA | AK101764.1 | - | 5 - 6 | 1660 | 90 | |
More Information
Set Membership: |
|

LOC4330553
Genomic Map