Assay Details
| Gene Symbol: LOC4330599 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015771433.2 | XP_015626919.1 | 8 - 9 | 1424 | 60 | |
| GenBank mRNA | AK070290.1 | - | 8 - 9 | 1400 | 60 | |
| AK289027.1 | - | 8 - 9 | 1418 | 60 | ||
More Information
Set Membership: |
|

LOC4330599
Genomic Map