Assay Details
| Gene Symbol: LOC4330604 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015770105.2 | XP_015625591.1 | 3 - 4 | 485 | 70 | |
| XM_015770106.2 | XP_015625592.1 | 2 - 3 | 521 | 70 | ||
| GenBank mRNA | AK109840.1 | - | 2 - 3 | 516 | 70 | |
More Information
Set Membership: |
|

LOC4330604
Genomic Map