Assay Details
| Gene Symbol: LOC4330648 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015767381.2 | XP_015622867.1 | 1 - 2 | 1046 | 66 | |
| GenBank mRNA | AK120606.1 | - | 1 - 2 | 1022 | 66 | |
More Information
Set Membership: |
|

LOC4330648
Genomic Map