Assay Details
| Gene Symbol: LOC4330648 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015767381.2 | XP_015622867.1 | 2 - 3 | 1195 | 65 | |
| GenBank mRNA | AK120606.1 | - | 2 - 3 | 1171 | 65 | |
More Information
Set Membership: |
|

LOC4330648
Genomic Map