Assay Details
| Gene Symbol: LOC4330732 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015771273.2 | XP_015626759.1 | 2 - 3 | 360 | 108 | |
| GenBank mRNA | AB188834.1 | - | 2 - 3 | 252 | 108 | |
| AK066687.1 | - | 2 - 3 | 332 | 108 | ||
More Information
Set Membership: |
|

LOC4330732
Genomic Map