Assay Details
| Gene Symbol: LOC4330732 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015771273.2 | XP_015626759.1 | 5 - 6 | 577 | 106 | |
| GenBank mRNA | AB188834.1 | - | 5 - 6 | 469 | 106 | |
| AK066687.1 | - | 5 - 6 | 549 | 106 | ||
More Information
Set Membership: |
|

LOC4330732
Genomic Map