Assay Details
| Gene Symbol: LOC4330732 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015771273.2 | XP_015626759.1 | 8 - 9 | 867 | 84 | |
| GenBank mRNA | AB188834.1 | - | 8 - 9 | 759 | 84 | |
| AK066687.1 | - | 8 - 9 | 1028 | 84 | ||
| BT130982.1 | - | 2 - 3 | 114 | 84 | ||
More Information
Set Membership: |
|

LOC4330732
Genomic Map