Assay Details
| Gene Symbol: LOC4330805 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015769429.2 | XP_015624915.1 | 7 - 8 | 958 | 100 | |
| GenBank mRNA | AB003322.1 | - | 7 - 8 | 758 | 100 | |
| AB107957.1 | - | 7 - 8 | 758 | 100 | ||
| AK070121.1 | - | 7 - 8 | 909 | 100 | ||
More Information
Set Membership: |
|

LOC4330805
Genomic Map