Assay Details
| Gene Symbol: LOC4331787 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015777261.2 | XP_015632747.1 | 3 - 4 | 853 | 91 | |
| XM_015777262.2 | XP_015632748.1 | 3 - 4 | 608 | 91 | ||
| GenBank mRNA | AK063978.1 | - | 3 - 4 | 511 | 91 | |
More Information
Set Membership: |
|

LOC4331787
Genomic Map