Assay Details
| Gene Symbol: LOC4331834 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015777661.2 | XP_015633147.1 | 1 | 852 | 106 | |
| GenBank mRNA | AK067971.1 | - | 1 | 766 | 106 | |
| AK120087.1 | - | 1 | 858 | 106 | ||
More Information
Set Membership: |
|

LOC4331834
Genomic Map