Assay Details
| Gene Symbol: LOC4332596 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015777067.2 | XP_015632553.1 | 4 - 5 | 966 | 109 | |
| GenBank mRNA | AB863631.1 | - | 4 - 5 | 853 | 109 | |
| AB863632.1 | - | 4 - 5 | 1549 | 109 | ||
| AK070233.1 | - | 1 - 2 | 88 | 109 | ||
| AK070638.1 | - | 4 - 5 | 939 | 109 | ||
More Information
Set Membership: |
|

LOC4332596
Genomic Map