Assay Details
| Gene Symbol: LOC4333649 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015777424.2 | XP_015632910.1 | 1 | 824 | 153 | |
| Gene Symbol: LOC4327264 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK240875.1 | - | 811 | 153 | ||
| AK287563.1 | - | 811 | 153 | |||
More Information
Set Membership: |
|

LOC4333649
Genomic Map