Assay Details
| Gene Symbol: LOC4333729 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015772905.2 | XP_015628391.1 | 5 - 6 | 574 | 83 | |
| XM_015772906.2 | XP_015628392.1 | 6 - 7 | 671 | 83 | ||
| GenBank mRNA | AK111767.1 | - | 5 - 6 | 583 | 83 | |
More Information
Set Membership: |
|

LOC4333729
Genomic Map