Assay Details
Gene Symbol: LOC4333729 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015772905.2 | XP_015628391.1 | 6 - 7 | 651 | 74 | |
XM_015772906.2 | XP_015628392.1 | 7 - 8 | 748 | 74 | ||
GenBank mRNA | AK111767.1 | - | 6 - 7 | 660 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
