Assay Details
| Gene Symbol: LOC4333742 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015777558.2 | XP_015633044.1 | 3 - 4 | 641 | 108 | |
| GenBank mRNA | AK288947.1 | - | 3 - 4 | 604 | 108 | |
More Information
Set Membership: |
|

LOC4333742
Genomic Map